Learn More
α-Cyclodextrin, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 10016-20-3 |
| Molecular Formula | C36H60O30 |
| MDL Number | MFCD00078207 |
| Quantity | 1 kg |
| SMILES | O[C@@H]1[C@H]([C@@H]2O[C@@H]3[C@@H]([C@H]([C@@H]([C@H](O3)CO)O[C@H](O[C@H]4CO)[C@@H](O)[C@H](O)[C@H]4O[C@H](O[C@H]5CO)[C@@H](O)[C@H](O)[C@H]5O[C@H](O[C@H]6CO)[C@@H](O)[C@H](O)[C@H]6O[C@@H](O[C@@H]7CO)[C@H](O)[C@@H](O)[C@@H]7O[C@H]1O[C@@H]2CO)O)O)O |
| Molecular Weight (g/mol) | 972.84 |
| Percent Purity | 97% |
| Chemical Name or Material | α-Cyclodextrin |