Learn More
Cyanuric Chloride, TRC
Product Code. 30009449
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009449 5 g 5g
30136935 500 g 500g
30003889 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009449 Supplier Toronto Research Chemicals Supplier No. TRCC9876955G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.41
Synonym 1,3,5-Trichloro-2,4,6-triazine, 1,3,5-Trichlorotriazine, 2,4,6-Trichloro-1,3,5-triazine, 2,4,6-Trichloro-s-triazine, 2,4,6-Trichloro-sym-triazine, 2,4,6-Trichlorotriazine, Cyanur Chloride, Cyanuric Chloride, Cyanuric Trichloride, Isocyanuric Trichloride, NSC 3512, Solgel W 08, Trichloro-s-triazine, Trichlorocyanidine, Zorugeru W 08, s-Triazine Trichloride, sym-Trichlorotriazine;
IUPAC Name 2,4,6-trichloro-1,3,5-triazine
SMILES Clc1nc(Cl)nc(Cl)n1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C3Cl3N3/c4-1-7-2(5)9-3(6)8-1
Formula Weight 182.9158
Chemical Name or Material Cyanuric Chloride