Learn More
Hanna Instruments™ Cyanuric Acid Reagent Water Testing Kit
Specifications
Specifications
| No. of Tests | 100 |
| Quantity | 100/Pk. |
| Range | 0 to 80mg/L |
| Test Method | Turbidimetric |
| Type | Cyanuric Acid Reagent |