Learn More
Hanna Instruments™ Cyanuric Acid Reagent Water Testing Kit
Product Code. 11863643
Change view
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11863643 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11863643 Supplier Hanna Instruments™ Supplier No. HI9372201
Specifications
No. of Tests 100
Quantity 100/Pk.
Range 0 to 80mg/L
Test Method Turbidimetric
Type Cyanuric Acid Reagent