Learn More
Macherey-Nagel™ Cyantesmo Qualitative Test Paper
Product Code. 11346504
Click to view available options
Color Indicator:
Pale Green to Blue
Unit Size:
Each
Product Code. 11346504
Brand: Macherey-Nagel™ 90604
Cyantesmo allows the quick and easy detection of hydrocyanic acid and cyanides in aqueous solutions and extracts.
Specifications
Color Indicator Pale Green to Blue
Format Paper
For Use With (Application) For detection of Hydrocyanic acid, Cyanides
Quantity 1 Each
Type Qualitative Test Paper
compliance-icons
Product Identifier
  • Cyantesmo
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : N,N,N',N'-tetramethyl-4,4'-diaminodiphenylmethane