missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cyanine 5, SE

Product Code. 30306096
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 30306096

Brand: Tocris Bioscience™ 5436/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Red fluorescent dye (Reactive group: NHS ester). Application: confocal microscopy, fluorescence microscopy, flow cytometry

Key information: Cyanine 5, SE is a widely used red fluorescent dye supplied with an NHS ester reactive group for the labeling of primary amines. Application: Suitable for fluorescence microscopy, confocal microscopy and flow cytometry. Properties and Photophysical Data: Excitation and emission maxima (λ) are 649 nm and 666 nm, respectively; quantum yield = 0.2; extinction coefficient = 250,000 M-1cm-1; A280 correction factor = 0.04. For more information on how Cyanine 5, SE may be used, see our protocol: Conjugation Protocol for Amine Reactive Dyes .
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[5-[1-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-1,3-dihydro-3,3-dimethyl-5-sulfo-2H-indol-2-ylidene]-1,3-pentadien-1-yl]-1-ethyl-3,3-dimethyl-5-sulfo-3H-indolium, inner salt
CAS 146368-14-1
Quantity 5 mg
Target Fluorescent Dyes
Molecular Formula C37H43N3O10S2
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.