Learn More
Cyanide, Quantofix™ Test Strips
Product Code. 15421127
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 15421127
Brand: Thermo Scientific Alfa Aesar 023273.QA
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
What instructions should I use for the Cyanide, Quantofix Test Strips?
We recommend the following instructions for the Cyanide, Quantofix Test Strips:
1. Rinse the measuring vessel with the test solution and fill it to the 5 mL mark.
2. Add 1 spoon of Cyanide-1 to the sample.
3. Add 5 drops of Cyanide-2 to the sample.
4. Shake the sample carefully.
5. Immediately dip the test strip into the sample.
6. Wait 45 seconds and shake off the excess liquid.
7. Compare within 10 seconds to the color scale.
Note: To obtain correct results with this test, the pH value of the sample must be <10. Therefore, the pH value of the sample must be measured prior to using the test strip. If the solution has a pH value >10, add drops of dilute sulfuric acid to lower the pH to 6-7 (optimal pH value).
What comes with the Cyanide, Quantofix Test Strips?
When you purchase the Cyanide, Quantofix Test Strips, you will receive the following:
  • 1 container with 100 test strips
  • 1 bottle of Cyanide-1
  • 1 bottle of Cyanide-2
  • 1 measuring vessel with 5 mL mark
  • 1 small measuring spoon
What are the graduations of the Cyanide, Quantofix Test Strips?
The graduations are as follows: 0 – 1 – 3 – 10 – 30 mg/L CN-