Learn More
Cyamemazine, TRC
Product Code. 17962525
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962525 10 mg 10mg
17954425 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17962525 Supplier Toronto Research Chemicals Supplier No. TRCC95150010MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3546-03-0
Molecular Formula C19 H21 N3 S
Molecular Weight (g/mol) 323.46
Synonym 10-[3-(Dimethylamino)-2-methylpropyl]-10H-phenothiazine-2-carbonitrile (ACI), Phenothiazine-2-carbonitrile, 10-[3-(dimethylamino)-2-methylpropyl]- (6CI, 7CI, 8CI), 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine-2-carbonitrile, 2-Cyano-10-(3-dimethylamino-2-methylpropyl)phenothiazine, 7204 R. E., 7204 RP, Ciamatil, Cianatil, Cyamemazin, Cyamemazine, Cyamepromazine, Cyamepromezine, F. I. 6229, Kyamepromazine, R.P. 7204, Tercian, TH 2602
IUPAC Name 10-[3-(dimethylamino)-2-methylpropyl]phenothiazine-2-carbonitrile
SMILES CC(CN(C)C)CN1c2ccccc2Sc3ccc(cc13)C#N
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C19H21N3S/c1-14(12-21(2)3)13-22-16-6-4-5-7-18(16)23-19-9-8-15(11-20)10-17(19)22/h4-10,14H,12-13H2,1-3H3
Formula Weight 323.1456
Percent Purity >95%
Chemical Name or Material Cyamemazine
Grade HPLC