missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Cy3.5 NHS Esters
Shop All Cytiva Products
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
The packs contain derivitised CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.
- Targeted and specific labeling of amine residues on antibodies, peptides or oligonucleotides.
- Bulk packs available for greatest economy.
- Example protocols included with each dye pack.
Specifications
Specifications
| Formulation | 100% Cy3.5 (NHS ester) |
| For Use With (Application) | For labelling via amine groups. CyDyes are suitable for use in applications such as fluorescence resonance energy transfer (FRET), time resolved- fluorescence resonance energy transfer (TR-FRET), and fluorescence polarization (FP) |
| Quantity | 25 mg |
| Product Type | Cy3.5 NHS Ester |
| Kit Contents | Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye |
| Molecular Weight (g/mol) | 1102.37 |
| Solubility | Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy3.5 |
| Color | Red |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction