Learn More
Cytiva Cy™5.5 Amidite
Product Code. 11530654 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11530654 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11530654 Supplier Cytiva Supplier No. 28904250
  • ONLINE EXCLUSIVE PRICE
    688.00€
    648.35€ / 100mg
    Save 39.65€
    5% Off
 Restricted Item
Estimated Shipment: 19-11-2026
For CyDye™ incorporation at any position in an oligonucleotide during synthesis
CyDye™ fluors combine exceptional photostability with bright and intense signals, resulting in high sensitivity and a highly reproducible performance at a biologically suitable pH range of 3 to 10.
  • Wide selection of CyDye™ fluors for protein labeling for use in ELISA & immunoprecipitation
  • Labeling strategy based on protein characteristics
  • Medium to high-throughput fluorescence imaging
  • Ready-to-use labeled antibodies enable multiplexed assays
  • Streptavidin-conjugated CyDye™ fluors for use in Flow-cytometry and In situ hybridization
  • Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
  • DNA synthesis reagent
Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye™ fluors enables multiplexing without crosstalk. In addition, the CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham™ CyDye™ fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials
Description Stable
Label or Dye Cy5.5
Product Line Cy
Product Type Cy5.5 Amidite
Quantity 100 mg
Size 100 mg
Form Solid