missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL17/VCC-1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF4207-SP
This item is not returnable.
View return policy
Description
CXCL17/VCC-1 Polyclonal specifically detects CXCL17/VCC-1 in Human samples. It is validated for Western Blot.Specifications
| CXCL17/VCC-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 284340 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q6UXB2 | |
| CXCL17 | |
| E. coli-derived recombinant human DMC Leu24-Leu119 Accession # Q6UXB2 | |
| 25 μg | |
| Primary | |
| Detects human DMC in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse DMC is observed. | |
| Human | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction