Learn More
Carbolite™ CWF 12/23 Laboratory Chamber Furnace
Product Code. 12029852
Change view
Click to view available options
Analysis Time:
Heat-up time: 45 min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analysis Time Unit Size
12029852 Heat-up time: 45 min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12029852 Supplier Carbolite™ Supplier No. CWF1223230SN+&023216P1
  • ONLINE EXCLUSIVE PRICE
    13255.00€
    10905.00€ / Pack of 1
    Save 2350.00€
    17% Off
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used as general purpose laboratory chamber furnaces with maximum temperature of 1200°C.
  • Maximum operating temperature: 1200°C
  • Parallel action door keeps heated surface away from the user
  • Delayed start/process timer function as standard
  • Hard wearing alumina element carrier, furnace entrance and hearth
  • Energy efficient low thermal mass insulation
  • Free radiating wire wound elements for optimum uniformity
  • Easy access to elements, controls simplifies maintenance and servicing
Specifications
Analysis Time Heat-up time: 45 min.
Capacity (Metric) 23 L
Temperature (Metric) 1200°C
Type Laboratory Chamber Furnace
Power Consumption 7000 w
Style Thermocouple Type: R
Dimensions (D x W x H) Interior 400 x 245 x 235 mm
Weight (Metric) 68
Wattage 2250 w holding