Learn More
CV 1808, Tocris Bioscience™
Product Code. 18095142
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
18095142 10 mg 10mg
1 options
Product Code. 18095142
Brand: Tocris Bioscience 1710/10
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Non-selective adenosine A2 receptor agonist
Adenosine A2 receptor agonist. Coronary vasodilator, antihypertensive and antipsychotic following systemic administration in vivo.
TRUSTED_SUSTAINABILITY
Specifications
CAS 53296-10-9
Molecular Formula C16H18N6O4
Quantity 10 mg
Synonym 2-phenylaminoadenosine, 2r,3r,4s,5r-2-6-amino-2-phenylamino-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, unii-k40i59g1ef, adenosine, 2-phenylamino, n6-phenylado, d06nrx, gtpl420, 3r,4s,5r-2-6-amino-2-anilino-purin-9-yl-5-hydroxymethyl oxolane-3,4-diol, 2-phenylamino ado cv-1808, 2-phenylaminoadenosine, solid
InChI Key SCNILGOVBBRMBK-SDBHATRESA-N
SMILES C1=CC=C(C=C1)NC2=NC3=C(C(=N2)N)N=CN3C4C(C(C(O4)CO)O)O
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-anilinopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 358.358
PubChem CID 6917803
Formula Weight 358.36
Percent Purity >99%
Chemical Name or Material CV 1808