Learn More
Hanna Instruments™ Cuvette Cleaning Solution
Product Code. 10608251
Change view
Click to view available options
Quantity:
230 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10608251 230 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10608251 Supplier Hanna Instruments™ Supplier No. HI9370350
  • ONLINE EXCLUSIVE PRICE
    94.69€
    54.71€ / Pack of 1
    Save 39.98€
    42% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Specifications
Color Clear to Pink
Composition Ethyl Alcohol
Concentration >15 to <35%
For Use With (Equipment) Photometers and Turbidity Meters
Filling Solution Type Buffer
Odor Alcohol-like
Optical Density 0.94 g/cm3
Packaging Bottle
pH Range 7
Quantity 230 mL
Solubility Water Soluble
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cuvette Cleaning Solution
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use powder to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol