Learn More
WTW™ Cuvette Cells for PhotoLab™ Photometers
Click to view available options
Path Length:
10 mm
50 mm
Unit Size:
Each
Description
- Cells with matt side surface are suitable for the automatic cell detection of the photoLab™ filter and spectral photometer
- All glass and quartz cuvettes have placement marks to locate them in the photometer in the same orientation for even higher precision
- High quality optical glass for VIS measurements, high quality quartz glass for UV-VIS
Specifications
Specifications
| Type | Cuvette |
| Description | REK 50 |
| Disposable | Reusable |
| Material | Quartz |
| Path Length | 50 mm |
| For Use With (Equipment) | Photometers |
| Shape | Rectangle |