Learn More
B Braun™ Cutfix Stainless Steel Scalpels
Product Code. 11995756
Change view
Click to view available options
Size:
# 10
# 11
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11995756 # 10 Pack of 10
10694161 # 11 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11995756 Supplier B Braun™ Supplier No. 5518059
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use B Braun™ Cutfix Sterile Stainless Steel Scalpels, with their double blade support, for precise cutting as your needs. This scalpel has a plastic handle.
  • Disposable
  • Sterile
  • Stainless steel
  • Two-sided blade support
Specifications
Blade Material Stainless Steel
For Use With (Application) Use for Dental Surgery
Size # 10
Disposable Single-use
Sterility Sterile
Handle Material Plastic