Learn More
3M™ Safety-Walk™ Cushion Matting 5270E
Product Code. p-7213716
Click to view available options
Length (Metric):
3.1 m
6.1 m
9.1 m
Unit Size:
1 roll
Each
Product Code. 17148740
Brand: 3M™ Safety-Walk™ 7100116724
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This motor oil resistant mat, suitable for heavy foot traffic in dry areas, provides a comfortable surface for walking or standing.
  • Coiled cushion matting compresses evenly across the bottom of the shoe, reducing leg and back fatigue
  • Textured top improves traction for safer footing
  • Suitable for dry work areas such as assembly lines and production areas
  • Supplied with edging to allow for wheeled traffic
  • Withstands most industrial and cleaning chemicals
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Length (English) 10 ft.
Length (Metric) 3.1 m
Type Cushion Mat
Width (English) 3 ft.
Width (Metric) 0.91 m