Learn More
Curdlan, TRC
Specifications
Specifications
| Chemical Name or Material | beta-1,3-Glucan |
| CAS | 54724-00-4 |
| Molecular Formula | C6H12O6 |
| Quantity | 1 g |
| Synonym | Biopoly P, Biopoly P 103, Curdlan NS, Curdran, β-1,3-Glucan Hydrate |
| InChI Formula | InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6-/m1/s1 |
| SMILES | C([C@@H]1[C@H]([C@@H]([C@H]([C@@H](O1)O)O)O)O)O |
| IUPAC Name | (2R,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| Molecular Weight (g/mol) | 180.16 |