missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Sulfate Pentahydrate, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Max % | 102 |
| Physical Form | Fine Crystals or Crystalline Powder |
| Quantity | 125 g |
| Percent Purity | 98 to 102% |
| Grade | ACS |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Cupric Sulfate Pentahydrate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?