Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
Product Code. 18779546
Change view
Click to view available options
Quantity:
1 mL
10 mL (2 x 5 mL)
5 mL
Unit Size:
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18779546 1 mL 1mL
18777896 5 mL 5mL
18737206 10 mL (2 x 5 mL) 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18779546 Supplier R&D Systems™ Supplier No. BME00101
  • ONLINE EXCLUSIVE PRICE
    108.00€
    101.85€ / 1mL
    Save 6.15€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-09-2026
Specifications
Quantity 1 mL
Species Mouse
Content And Storage Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.