Learn More
R&D Systems™ Cultrex UltiMatrix RGF Basement Membrane Extract
Product Code. 18777896
Change view
Click to view available options
Quantity:
1 mL
10 mL (2 x 5 mL)
5 mL
Unit Size:
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777896 5 mL 5mL
18779546 1 mL 1mL
18737206 10 mL (2 x 5 mL) 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18777896 Supplier R&D Systems™ Supplier No. BME00105
  • ONLINE EXCLUSIVE PRICE
    437.00€
    412.65€ / 5mL
    Save 24.35€
    5% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Specifications
Product Line R&D Systems
Quantity 5 mL
Species Mouse
Content And Storage Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.