Learn More
Thermo Scientific™ Culti-Loops™ Beta-lactamase Test QC Set
Product Code. 15269277
Click to view available options
Quantity:
5 Loops/Pack
Unit Size:
Each
Product Code. 15269277
Brand: Thermo Scientific™ R4651109
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ Beta-lactamase Test Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Neisseria gonorrhoeae ATCC™ 31426™†
Neisseria gonorrhoeae ATCC™ 43069™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 31426TM*
Description CLIA '88 Compliance Beta-lactamase Test QC Set/2
Format Culti-Loops
Product Type Culti-Loops Quality Control Organism
Target Organism Class Organism Sets
Test Type Qualitative QC
Quantity 5 Loops/Pack
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.