Learn More
Thermo Scientific™ Culti-Loops™ API™ 20E QC Set
Product Code. 15209277
Click to view available options
Quantity:
5 Loops/Organism
Unit Size:
Each
Product Code. 15209277
Brand: Thermo Scientific™ R4651003
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ API 20E Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Enterobacter cloacae ATCC™ 13047™†
Escherichia coli ATCC™ 25922™†
Klebsiella pneumoniae subsp. pneumoniae ATCC™ 35657™†
Proteus mirabilis ATCC™ 35659™†
Stenotrophomonas maltophilia ATCC™ 51331™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 13047TM*
Description API 20E QC Set/5
Format Culti-Loops
Product Type Culti-Loops QC Set
Target Organism Class Organism Sets
Test Type Qualitative QC
Quantity 5 Loops/Organism
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.