Learn More
Gibco™ CTS™ ACK Lysing Buffer
Description
Gibco Cell Therapy Systems (CTS) ACK (Ammonium-Chloride-Potassium) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow. The complete formulation and lysis protocol are available.
CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.
Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.
Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.
Specifications
Specifications
| Content And Storage | 15°C to 30°CC. Protect from light. |
| Culture Type | Mammalian cell culture |
| Form | Clear, colorless solution |
| Product Type | Cell Culture Supplement |
| Sterility | Sterile-filtered |
| Sterilization Method | 0.2 μm Sterile-filtered |
| Format | Bottle |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Manufacturing Quality | ISO13485, MDSAP, FDA-registered, 21 CFR820 |
| Quantity | 100 mL |
| Show More |
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.