Learn More
Kimble Chase™ CTFE/Stainless Steel Piston Pellet for Tissue Grinders
Click to view available options
Description:
Reusable PELLET PESTLE
Unit Size:
Each
Specifications
Specifications
| Description | Reusable PELLET PESTLE |
| Capacity (Metric) | 50 mL |
| Autoclavable | Autoclavable |
| Product Type | Pestle |
| Material | PTFE |
| For Use With (Equipment) | CTFE/Stainless Steel Tissue Grinders |
| Length (Metric) | 210mm |
| Type | Pestle for Tissue Grinder |