missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | CSN1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838973
|
Proteintech
11709-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16828973
|
Proteintech
11709-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
OTUD4, also known as HIV-1 induced protein HIN-1, is a member of the OTU (ovarian tumor) domain containing cysteine protease superfamily, in which the OUT domain generally confers deubiquitinase activity. At least three isoforms of OTUD4 are known to exist, and the smallest of these isoforms are only expressed in HIV-1-infected cells.Specifications
CSN1 | |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
Q13098 | |
2873 | |
CSN1 Fusion Protein Ag2273 | |
Primary | |
-20°C | |
GPS1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COPS1, CSN1, G protein pathway suppressor 1, GPS 1, GPS1, Protein MFH, SGN1, Signalosome subunit 1 | |
GPS1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |