missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-ADE-HIS-LEU-TRP, Powder

Product Code. 11337939 Shop All MP Biomedical Products
Change view
Click to view available options
Complete Supplement Mixture;
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11337939 Complete Supplement Mixture;
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11337939 Supplier MP Biomedicals™ Supplier No. 114540422
  • ONLINE EXCLUSIVE PRICE
    898.00€
    854.70€ / 100g
    Save 43.30€
    4% Off
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MP Biologicals™ 4 dropout formulation (without Adenine, Histidine, Leucine, and Tryptophan) of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiae growth media.

  • 4 dropout formulation of Complete Supplement Mixture (CSM) of amino acids for S. cerevisiae growth media.
  • Time tested formulation is available in complete and dropout formulations.
  • The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component.
  • This CSM-Dropout medium create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Yeast Supplement
Sterilization Method Autoclave at 121°C for 15 min.
pH 4.0 to 4.8 (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.