Learn More
BD Biosciences™ CS&T Research Beads for use with BD FACSDiva™ Version 7 or Later (RUO)
Click to view available options
No. of Tests:
150 Tests
50 Tests
Unit Size:
Each
Description
- Designed for use on BD™ flow cytometers running BD FACSuite™ software version 7.0 or later.
- FC beads enable the software to automatically characterize, track, and report measurements of supported BD digital flow cytometers.
- CS&T research beads are dyed with fluorochromes that are excited by the cytometer's lasers.
- Provides cost efficiency with low per-test cost and multiple kit configurations.
- Produced under GMP for improved quality and reproducibility.
- Enhances the cytometer setup workflow, supporting over 100 laser and filter combinations.
Specifications
Specifications
| Content And Storage | Three 3 mL vial at 50 tests per vial for 150 tests per kit |
| No. of Tests | 150 Tests |
| Stability | The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 24 hours at 2°C–8°CC or 8 hours at 15°C–25°C when protected from light |
| Product Type | Fluorescent Polystyrene Beads |