Learn More
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb
Description
- Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
- Superior chemical resistance compared to polycarbonate.
- Sterilize by autoclave, gamma irradiation or ethylene oxide.
- Meets USP Class VI.
- Animal-derived ingredient-free.
- Available in packs of 25.
Specifications
Specifications
| Description | Male luer lock |
| Material | Polyolefin |
| Packaging Quantity | 25/Pk. |
| Color | Berry |
| Product Type | Male Luer Lock Fitting |