Learn More
Multiroir™ Transparent Crystal Polystyrene Storage Drawers
Click to view available options
Capacity (Metric):
10 L
30 L
Unit Size:
Each
Description
- Total visibility
- Crystal polystyrene transparent - like glass
- Slide assembly
- Drawer separators available separately
Specifications
Specifications
| Capacity (Metric) | 10 L |
| Color | Transparent |
| Material | Polystyrene |
| Shape | Rectangular |
| Product Type | Storage Drawer |
| Quantity | 1 Each |
| Dimensions (L x W x H) | 410 x 265 x 125 mm |
| Length (Metric) | 410 mm |
| Length (Metric) Interior | 365 mm |
| Width (Metric) | 265 mm |
| Show More |