Learn More
STEMCELL Technologies CryoStor™ CS5
Product Code. 17149931
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
100 mL Bag
5 x 10 mL
Unit Size:
100mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17149931 100 mL Bag 100mL
17119931 5 x 10 mL Pack of 5
17129921 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17149931 Supplier STEMCELL Technologies Supplier No. 07953
 Restricted Item
Estimated Shipment: 04-11-2026
Animal-component-free, defined cryopreservation medium with 5% DMSO
  • CryoStor™ CS5 is a uniquely formulated serum-free, animal component-free and defined cryopreservation medium containing 5% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS5 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS5 is cGMP-manufactured with USP grade components
Specifications
Cell Type CHO Cells, Mesenchymal Stem and Progenitor Cells, Other, Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 100 mL Bag