Learn More
STEMCELL Technologies CryoStor™ CS2
Product Code. 17119921
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17119921 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17119921 Supplier STEMCELL Technologies Supplier No. 07932
 Restricted Item
Estimated Shipment: 08-10-2026
Animal component-free, defined cryopreservation medium with 2% DMSO
  • CryoStor™ CS2 is a uniquely formulated serum-free, animal component-free, and defined cryopreservation medium containing 2% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS2 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS2 is cGMP-manufactured with USP grade components
Specifications
Cell Type Mesenchymal Stem and Progenitor Cells; Other; Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 100 mL