Learn More
Thermo Scientific™ CryoMed™ Sensors
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
561.00€ - 594.00€
Specifications
Description | Thermocouple Sensor |
---|---|
Type | Thermocouple Sensor |
Plug Type | Male |
Holds | Cryo Tube, Sheath Straw, Vial |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10061204
|
Thermo Scientific™
4000385 |
1.2/2.0 Cryo Tubes in CryoMed |
594.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
10590275
|
Thermo Scientific™
4000386 |
4.0/5.0 Cryo Tubes in CryoMed |
561.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:- Thermocouple sensors for cryo tubes, sheath straw and vial samples
- Includes male plug
Recommended for:
Controlled-rate freezing applications
Specifications
Thermocouple Sensor | |
Thermocouple Sensor | |
Male | |
Cryo Tube, Sheath Straw, Vial |