Learn More
GA International Cryogenic Writable Labels
Product Code. 17879357
Encompass_Preferred
Click to view available options
Color:
Beige
Blue
Brown
Green
Lavender
Lemon
Mint
Orange
Pink
Seafoam
Silver
Violet
White
Unit Size:
Pack of 1000
Product Code. 17879357
Brand: GA International JTRA2610SI
This item requires a price update prior to being ordered.
Request a Price for this item here.
Writable cryogenic color rectangles provided in dispenser box rolls for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen Dewars.
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage in -80°C freezers, liquid nitrogen and dry ice
  • Available in a wide range of colors, for color coding and easy identification
  • Small round labels are ideal for Eppendorf tube caps and vials
  • Available in a variety of colors
  • Writable with permanent cryo markers
Specifications
Adhesive Type Permanent
Color Silver
Finish Matte
Format Roll
Material Thermoplastic Film
Product Type Label
Sterility Non-sterile
Labels per Row 1 Label
Quantity 1000/Pk.
Shape Rectangle
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Dimensions (LxW) 26 x 10 mm
Length (English) 0.4 in.
Length (Metric) 10 mm
Width (English) 1.02 in.
Width (Metric) 26 mm
Diameter (English) 1 in.
Diameter (Metric) 25.4 mm
Show More Show Less
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.