Learn More
BRAND™ Cryogenic Tubes Storage Box
Shop All Brand Products
Click to view available options
Holds:
100 Tubes
Unit Size:
Pack of 4
Description
- For cryogenic tubes
- Operating range (in the gaseous phase of liquid nitrogen) between -196 to 121°C
- Autoclavable at 121°C (2 bar), acc. DIN EN 285
Specifications
Specifications
| Material | Polycarbonate |
| Type | Storage Box |
| For Use With (Equipment) | 1.2 mL Cryogenic Tubes, 2 mL Cryogenic Tubes |
| Holds | 100 Tubes |
| Length (Metric) | 20.9 cm |
| Width (Metric) | 13 cm |
| Height (Metric) | 13 cm |
| Weight (Metric) | 677.9 g |