Learn More
Cole Parmer Cryogenic Labels
Cole Parmer Cryogenic Labels are for use with laser printers. The heat resistant, stay-flat sheets will not jam in laser printers.
Brand:  Cole Parmer 62206-70
132.05 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 1.00000kg
Product Code. 12867690
  • 139.00€ / Pack of 1700
Estimated Shipment: 09-07-2024
Description
Cole Parmer Cryogenic Labels are for use with laser printers. The heat resistant, stay-flat sheets will not jam in laser printers.
Specifications
Specifications
Cold Temperature
1-9/32 x 1/2 in.
0.5 in.
Laser Printers
-190°C to +150°C