Learn More
Sperian™ Honeywell™ Cryogenic Gloves
Click to view available options
Size:
9
10
11
Unit Size:
Each
Description
- Water-repellant silicone cowhide grain leather
- Gunn cut, winged thumb, crispin style
- Reinforcement between thumb and index finger
- 200mm split leather cuff
- Velcro™ adjustment strap on side of the palm
- Cuff offers good forearm protection against mechanical hazards and limits any risk of intrusion
Specifications
Specifications
| Size | 10 |
| Length (Metric) | 400 mm |