Learn More
Ansell Edmont™ Crusader Flex™ 42-445 Series Heavy Duty Nitrile Gloves
Shop All Ansell Products
Click to view available options
Size:
8
Unit Size:
12 pairs
Description
- Provides high resistance to cuts, minor injuries and abrasion
- Enhances grip on dry and oily surfaces
- Comfortable and flexible: sweat absorbent non-woven felt inner
Specifications
Specifications
| Certifications/Compliance | EN 420:2003 + A1:2009 and EN 388:2003 , EN 407:2004 |
| Description | Construction: Cut and sewn |
| Coating Material | Uncoated |
| Color | Gray |
| Exterior Finish | No Texture |
| Hand | Specific |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Heavy-duty Nitrile Gloves |
| Size | 8 |
| Show More |