Learn More
Crison CRISOLYT-A electrolytic solution
Click to view available options
Composition:
3M KCl + AgCl
Unit Size:
Each
Specifications
Specifications
| Composition | 3M KCl + AgCl |
| Type | Electrolyte Solution |
| Packaging | Bottle |
| Quantity | 125 mL |