Learn More
Fisherbrand™ Crimping Heads for Programmable Electronic High Power Crimp Station
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Cap:
8 mm
11 mm
13 mm
20 mm
Unit Size:
Each
Specifications
Specifications
| Description | 13mm Crimping Head |
| Diameter (Metric) Cap | 13 mm |
| Material | Plastic, Steel |
| Product Type | Crimping Head |