Learn More
Creatine phosphate disodium salt tetrahydrate, 98+%
Product Code. 11448296
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448296 5 g 5g
11438296 1 g 1g
11458296 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11448296 Supplier Thermo Scientific Alfa Aesar Supplier No. A15362.06
 Restricted Item
In Stock
Add to basket
CAS: 71519-72-7 | C4H8N3Na2O5P·4H2O
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
Solubility
Soluble in water.0.1 g/mL.
Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71519-72-7
Molecular Formula C4H8N3Na2O5P·4H2O
MDL Number MFCD00150192
Synonym Sodium creatine phosphate dibasic tetrahydrate
Specifications
Assay Percent Range ≥98%
Quantity 5 g
Solubility Information Soluble in water.0.1g/mL.
Formula Weight 327.14 (255.07 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Creatine phosphate disodium salt tetrahydrate