Learn More
CPN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA596846
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: FVLSANLHGG AVVANYPYDK SFEHRVRGVR RTASTPTPDD KLFQKLAKVY SYAHGWMFQG WNCGDYFPDG ITNGASWYSL SKGMQDFNYL HTNCFEITLE LSCDKFPPEE ELQREWLGNR EALIQFLEQV HQGIKGMVLD ENYNNLANAV ISVSGINHDV TSGDHGDYFR LLLPGIYTVS ATAPGYDPET VTVTVGPAEP TLVNFHLKRS IPQVSPVRRA PSRRHGVRAK VQPQARKKEM EMRQLQRGPA; Positive Samples: MCF-7, Raji, Mouse liver, Mouse pancreas, Rat brain; Cellular Location: extracellular space
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.Specifications
CPN1 | |
Polyclonal | |
Unconjugated | |
CPN1 | |
CPN1 | |
Recombit fusion protein containing a sequence corresponding to amino acids 209-458 of human CPN1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
4.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15169, Q9EQV8, Q9JJN5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1369, 365466, 93721 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |