Learn More
Tocris Bioscience™ CPI 203
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
CPI 203 is a BET bromodomain inhibitor. Downregulates Myc expression, causes G1 cell cycle arrest and attenuates cell proliferation in human pancreatic neuroendocrine tumors. Arrests the growth of T cell acute lymphoblastic leukemia cells in vitro (EC50 = 91.2 nM). Also enhances the antitumor effect of rapamycin (Cat. No. 1292) in vitro and attenuates rapamycin induced Akt activation. Also enhances fibroblast reprogramming to hiPSCs at low concentration. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | CPI 203 |
| Purity | >98% |
| Molecular Weight (g/mol) | 399.9 |
| Inhibitors | CPI 203 |