missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CP 465022 hydrochloride
GREENER_CHOICE

Product Code. 18004712
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004712

Brand: Tocris Bioscience™ 2932/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective, non-competitive AMPA antagonist

Selective, non-competitive AMPA antagonist (IC50 = 25 nM in rat cortical neurons) that displays potent anticonvulsant activity. Also significantly blocks the persistent component of Nav1.6 channel activity. Brain penetrant and orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1785666-59-2
Molecular Formula C26H25Cl2FN4O

Specifications

Quantity 50 mg
Formula Weight 499.41
Purity >99%
Molecular Weight (g/mol) 499.411
Chemical Name or Material CP 465022 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.