Learn More
Sperian™ Cowhide Leather Gloves
Click to view available options
:
Size: 8 to 9 to 10 to 11
Specifications
Specifications
| Certifications/Compliance | EN 420 / EN 388 / EN 407 |
| Cuff Material | Cotton |
| Cut Resistance Rating | 1 |
| Material | Cowhide Leather |
| Product Type | Gloves |
| Size | 8, 9, 10, 11 |
| Thickness (Metric) | 1.3 mm |
| Lining | Cotton |
| Model | T08 |
| Resistant To | Heat |
| Show More |