Learn More
KARTELL Cover for Microtitre Plates
Product Code. 12860923
Change view
Click to view available options
Plate Compatibility:
Plate Codes 2620, 2621, 2622
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plate Compatibility Unit Size
12860923 Plate Codes 2620, 2621, 2622 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12860923 Supplier KARTELL Supplier No. 02623/S
  • ONLINE EXCLUSIVE PRICE
    40.00€
    37.73€ / Pack of 50
    Save 2.27€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-01-2027
These Microtitre Plate Covers are compatible with flat-bottom, U-bottom and V-bottom well plates.
  • Sterile PS covers for Kartell Microtitrations plates 0262000, 0262200 and 262100
  • Available also in non sterile version (code 0262300).
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Adhesive None
Plate Compatibility Plate Codes 2620, 2621, 2622
Fit for Purpose Sterile