Learn More
Macherey-Nagel™ Cover Lids for NucleoFast™ 96 PCR Plates
Product Code. 15850978
Change view
Click to view available options
:
50 Lids
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15850978 50 Lids
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15850978 Supplier Macherey-Nagel™ Supplier No. 743101.50
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
To prevent contamination and spills when working with NucleoFast™ 96-well ultrafiltration plates for PCR clean-up.
NucleoFast™ 96 PCR
  • Cost-efficient 96-well ultrafiltration kit for PCR clean-up
  • Ready-to-use DNA for sequencing and microarray spotting
  • Sturdy membrane allows easy recovery of DNA
  • No well-to-well cross-contamination
  • Fast procedure
  • Available as complete kits and as plates only
  • Detergent-free membrane
  • Manual or automated use