Learn More
Marienfeld Superior™ Cover Glasses for Counting Chambers and Haemacytometers
Product Code. p-7144731
Click to view available options
Dimensions (L x W):
24 x 24 mm
Unit Size:
Pack of 100
Product Code. 10147771
Brand: Marienfeld Superior™ 0352000
Use Marienfeld Superior™ Cover Glasses of 0.4 mm thickness for counting chambers and haemacytometers for covering the specimens in counting chambers.
  • Made of special optical glass
  • Ready for use
  • Ground and polished edges
  • Additional feature: The excellent quality of the edges make the haemacytometer cover glasses well suitable for blood smears
  • Individually packed in poly bags, 10 poly bags in a plastic box, 10 plastic boxes in a cardboard box
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark and batch number for comprehensive information and traceability
Specifications
Product Type Cover Glass
Shape Square
Material Special Optical Glass
Thickness (Metric) 0.4 mm
Dimensions (L x W) 24 x 24 mm
For Use With (Equipment) Counting Chambers and Haemacytometers
Quantity 100