Learn More
Hirschmann™ Cover Glasses for Microscopy
Product Code. p-8011834
Change view
Click to view available options
Dimensions (L x W):
15 x 15 mm
18 x 18 mm
20 x 20 mm
22 x 22 mm
22 x 32 mm
22 x 40 mm
22 x 50 mm
22 x 60 mm
24 x 24 mm
24 x 32 mm
24 x 46 mm
24 x 48 mm
24 x 50 mm
Unit Size:
Pack of 1000
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10062481 18 x 18 mm Pack of 1000
10650193 24 x 24 mm Pack of 1000
13168159 22 x 40 mm Pack of 1000
13128159 22 x 22 mm Pack of 1000
13198159 24 x 32 mm Pack of 1000
13230480 24 x 50 mm Pack of 1000
13158159 22 x 32 mm Pack of 1000
10531942 20 x 20 mm Pack of 1000
13178159 22 x 50 mm Pack of 1000
10165831 15 x 15 mm Pack of 1000
13118169 24 x 46 mm Pack of 1000
13128169 24 x 48 mm Pack of 1000
13188159 22 x 60 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 10062481 Supplier Hirschmann™ Supplier No. 8000102
Preserve and protect specimens on slides. Hirschmann Instruments™ Cover Glasses for Microscopy are manufactured from quality glass and come in a range of dimensions.
  • 0.13-0.17 mm thickness
  • Acc. to DIN 58 884, CE acc. IVD 98/79 EC
  • Provided in packs of 1000
  • With lot number on packaging
Specifications
Product Type Coverslip
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 18 x 18 mm
Certifications CE, DIN 58884, IVD 98/79 EC