Learn More
Carl Zeiss™ Cover Glasses, High Performance
Product Code. 10474379
Change view
Click to view available options
Diameter (Metric):
0.17 mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10474379 0.17 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10474379 Supplier Carl Zeiss™ Supplier No. 4740309000000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Recommended for applications with high numerical aperture objectives. Carl Zeiss™ Cover Glasses, High Performance come in a box of 100pcs.
  • Type 1 1/2 H as per ISO 8255-1
  • Restricted thickness-related tolerance D=0.17 mm +/- 0.005 mm
  • Refractive index = 1.5255 +/- 0.0015
  • Abbe number = 56 +/-2
Specifications
Product Type Coverslip
Diameter (Metric) 0.17 mm
Dimensions (L x W) 18 x 18 mm
Quantity 1000/Pk.