Learn More
Fisherbrand™ Cover for General Purpose Water Bath
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Precision 10L Water Baths
Precision 2L and 5L Water Baths
Unit Size:
Each
Description
Gassing Hoods
- Permit control over the atmosphere surrounding the sample
- Improve temperature uniformity and reduce energy consumption
- Polished stainless steel with plastic gas fitting
- Each bath will accommodate 1 large hood or 2 small hoods
- Enhanced hinged design supports open lid configuration and easy lid removal
- Accommodate taller labware with extended height
- Polycarbonate and stainless steel models available
Specifications
Specifications
| Description | Stainless Steel Gable Cover |
| For Use With (Equipment) | Precision 10L Water Baths |
| Type | Gable Cover |
| Material | Stainless Steel |